ENA238713854 Enamine

3-(3-aminophenoxy)propanamide

  • MDL number MFCD11206020
  • Cas Number : 1094322-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52208
Availability details for
Quantity
  • Availability
  • Add to Cart