ENA238713861 Enamine

1-(3,3-dimethylbutanoyl)piperidin-4-one

  • MDL number MFCD09929542
  • Cas Number : 1016705-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52211
Availability details for
Quantity
  • Availability
  • Add to Cart