ENA238713871 Enamine

1-benzoyl-1,4-diazepane

  • MDL number MFCD05879210
  • Cas Number : 59939-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52217
Availability details for
Quantity
  • Availability
  • Add to Cart