ENA238713875 Enamine

1-(1,4-diazepan-1-yl)butan-1-one

  • MDL number MFCD09811954
  • Cas Number : 61903-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52223

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52223
Availability details for
Quantity
  • Availability
  • Add to Cart