ENA238713880 Enamine

3-chloro-4-(2-methoxyethoxy)aniline

  • MDL number MFCD08056136
  • Cas Number : 879047-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-52226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52226
Availability details for
Quantity
  • Availability
  • Add to Cart