ENA245485092 Enamine

2-chloro-N-(2-cyanoethyl)-N-(4-methylphenyl)propanamide

  • MDL number MFCD13195800
  • Cas Number : 1210020-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28137
Availability details for
Quantity
  • Availability
  • Add to Cart