ENA245485105 Enamine

2-chloro-N-(1-propyl-1H-pyrazol-5-yl)acetamide

  • MDL number MFCD13195818
  • Cas Number : 1209931-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46661
Availability details for
Quantity
  • Availability
  • Add to Cart