ENA245485122 Enamine

2,2,2-trifluoroethyl N-[5-(propan-2-yl)-1,3,4-thiadiazol-2-yl]carbamate

  • MDL number MFCD13195857
  • Cas Number : 1198062-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52883
Availability details for
Quantity
  • Availability
  • Add to Cart