ENA245485123 Enamine

methyl 1,1-dioxo-3-phenyl-4H-1lambda6,2,4-benzothiadiazine-7-carboxylate

  • MDL number MFCD13195862
  • Cas Number : 1210774-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53095
Availability details for
Quantity
  • Availability
  • Add to Cart