ENA245485143 Enamine

N-butyl-2-chloropropanamide

  • MDL number MFCD09949372
  • Cas Number : 94318-73-7
  • Molecular Weight : 163.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27901
Availability details for
Quantity
  • Availability
  • Add to Cart