ENA245485145 Enamine

2-chloro-N-(3-methoxypropyl)propanamide

  • MDL number MFCD11205017
  • Cas Number : 1094292-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27905
Availability details for
Quantity
  • Availability
  • Add to Cart