ENA245485150 Enamine

2-chloro-N-(2-methylbutan-2-yl)propanamide

  • MDL number MFCD09930823
  • Cas Number : 42276-10-8
  • Molecular Weight : 177.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27941

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27941
Availability details for
Quantity
  • Availability
  • Add to Cart