ENA245485152 Enamine

2-chloro-N-(2-fluoro-4-methylphenyl)propanamide

  • MDL number MFCD11183594
  • Cas Number : 1039986-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27945

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27945
Availability details for
Quantity
  • Availability
  • Add to Cart