ENA245485157 Enamine

2-mercapto-3-[4-(trifluoromethoxy)phenyl]-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD06655459
  • Cas Number : 901272-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12734

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12734
Availability details for
Quantity
  • Availability
  • Add to Cart