ENA245485159 Enamine

3-(2-chloropropanamido)-N,N-diethylbenzamide

  • MDL number MFCD12655062
  • Cas Number : 1098347-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28047
Availability details for
Quantity
  • Availability
  • Add to Cart