ENA245485163 Enamine

2-chloro-1-(4-acetylpiperazin-1-yl)propan-1-one

  • MDL number MFCD09931241
  • Cas Number : 1016699-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28106

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28106
Availability details for
Quantity
  • Availability
  • Add to Cart