ENA245485171 Enamine

1-(4-aminophenyl)-3-[(oxolan-2-yl)methyl]urea

  • MDL number MFCD12412719
  • Cas Number : 1156147-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28230
Availability details for
Quantity
  • Availability
  • Add to Cart