ENA245485178 Enamine

methyl 2,4-dichloro-5-(chlorosulfonyl)benzoate

  • MDL number MFCD11099042
  • Cas Number : 924867-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28659

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28659
Availability details for
Quantity
  • Availability
  • Add to Cart