ENA245485184 Enamine

1-(2,4-dimethylphenyl)imidazolidine-2,4,5-trione

  • MDL number MFCD11205061
  • Cas Number : 1094424-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-29238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29238
Availability details for
Quantity
  • Availability
  • Add to Cart