ENA245485205 Enamine

N-ethyl-3-methoxyaniline

  • MDL number MFCD03210693
  • Cas Number : 41115-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33036

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33036
Availability details for
Quantity
  • Availability
  • Add to Cart