ENA245485220 Enamine

2-methyl-3-(1H-1,2,3,4-tetrazol-1-yl)aniline

  • MDL number MFCD06335065
  • Cas Number : 750599-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35279

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35279
Availability details for
Quantity
  • Availability
  • Add to Cart