ENA245485221 Enamine

2-[1-(difluoromethyl)-1H-1,3-benzodiazol-2-yl]propan-1-amine dihydrochloride

  • MDL number MFCD09285092
  • Cas Number : 1049702-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35364
Availability details for
Quantity
  • Availability
  • Add to Cart