ENA245485248 Enamine

1-(4-nitrophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08696935
  • Cas Number : 19532-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42129
Availability details for
Quantity
  • Availability
  • Add to Cart