ENA245485253 Enamine

6-methyl-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole

  • MDL number MFCD11212307
  • Cas Number : 139774-14-4
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methyl-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole
Inchi Code 1S/C14H16N2/c1-10-2-3-12-13(9-16-14(12)8-10)11-4-6-15-7-5-11/h2-4,8-9,15-16H,5-7H2,1H3
Inchi Key GCUXMIDTUTXWHW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-42506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42506
Availability details for
Quantity
  • Availability
  • Add to Cart