ENA245485266 Enamine

N'-hydroxycyclohexanecarboximidamide

  • MDL number MFCD09959849
  • Cas Number : 633313-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-45421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45421
Availability details for
Quantity
  • Availability
  • Add to Cart