ENA245485270 Enamine

2-chloro-N-(4-cyano-1-phenyl-1H-pyrazol-5-yl)acetamide

  • MDL number MFCD01989752
  • Cas Number : 397860-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46061
Availability details for
Quantity
  • Availability
  • Add to Cart