ENA245485306 Enamine

1-(4-cyanophenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD07364795
  • Cas Number : 944643-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50495

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50495
Availability details for
Quantity
  • Availability
  • Add to Cart