ENA245485310 Enamine

2-(adamantan-1-yl)-2-aminoacetic acid

  • MDL number MFCD01838120
  • Cas Number : 60256-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50676

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50676
Availability details for
Quantity
  • Availability
  • Add to Cart