ENA245485318 Enamine

3-(propan-2-yl)thiophene-2-carboxylic acid

  • MDL number MFCD12912914
  • Cas Number : 29488-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-110
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51047
Availability details for
Quantity
  • Availability
  • Add to Cart