ENA245485320 Enamine

2-{5-[(dimethylamino)methyl]-1,2,4-oxadiazol-3-yl}propan-2-amine dihydrochloride

  • MDL number MFCD13195827
  • Cas Number : 1209883-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51063
Availability details for
Quantity
  • Availability
  • Add to Cart