ENA245485323 Enamine

3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD00020813
  • Cas Number : 23598-72-3
  • Molecular Weight : 237.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193-194
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18171

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
MSDS https://www.enaminestore.com/catalog/EN300-18171
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart