ENA245485328 Enamine

5-chloro-[1,2,3,4]tetrazolo[1,5-a]pyrazine

  • MDL number MFCD13195832
  • Cas Number : 1210210-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51229
Availability details for
Quantity
  • Availability
  • Add to Cart