ENA245485335 Enamine

ethyl 2-(cyanomethyl)-1,3-thiazole-4-carboxylate

  • MDL number MFCD03294532
  • Cas Number : 59142-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51449
Availability details for
Quantity
  • Availability
  • Add to Cart