ENA245485343 Enamine

1-cyclopropylimidazolidine-2,4-dione

  • MDL number MFCD12159774
  • Cas Number : 1178392-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51672

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51672
Availability details for
Quantity
  • Availability
  • Add to Cart