ENA245485348 Enamine

2-[1-(3-chlorophenyl)-5-methyl-1H-1,2,3-triazol-4-yl]acetic acid

  • MDL number MFCD10006709
  • Cas Number : 1017457-61-2
  • Molecular Weight : 251.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51786
Availability details for
Quantity
  • Availability
  • Add to Cart