ENA245485366 Enamine

1-(2-aminophenyl)ethan-1-ol

  • MDL number MFCD12198838
  • Cas Number : 10517-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52136

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52136
Availability details for
Quantity
  • Availability
  • Add to Cart