ENA245485374 Enamine

2-(2-aminoethyl)-2,3-dihydro-1H-isoindole-1,3-dione hydrochloride

  • MDL number MFCD08272836
  • Cas Number : 30250-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 276-280
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52248

Safety Information

signalword Danger
pcodes P280,P305+P351+P338,P310
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-52248
pictograms GHS05
hstatements H318
Availability details for
Quantity
  • Availability
  • Add to Cart