ENA245485393 Enamine

2-butyl-1H-1,3-benzodiazol-5-amine dihydrochloride

  • MDL number MFCD12913107
  • Cas Number : 1197821-90-1
  • Molecular Weight : 262.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-52412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52412
Availability details for
Quantity
  • Availability
  • Add to Cart