ENA245485400 Enamine

methyl 2-(chlorosulfonyl)propanoate

  • MDL number MFCD11131128
  • Cas Number : 20449-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52458

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-52458
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart