ENA245485406 Enamine

2-methyl-2-{[(prop-2-en-1-yl)carbamoyl]amino}propanoic acid

  • MDL number MFCD09941258
  • Cas Number : 1016823-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52484
Availability details for
Quantity
  • Availability
  • Add to Cart