ENA245485414 Enamine

4-(3-fluorophenyl)butan-2-one

  • MDL number MFCD11553463
  • Cas Number : 3506-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52536

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52536
Availability details for
Quantity
  • Availability
  • Add to Cart