ENA245485423 Enamine

benzyl(oxolan-2-ylmethyl)amine

  • MDL number MFCD03001392
  • Cas Number : 183275-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52573

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52573
Availability details for
Quantity
  • Availability
  • Add to Cart