ENA245485433 Enamine

5-[4-(trifluoromethyl)phenyl]furan-2-carbaldehyde

  • MDL number MFCD06740260
  • Cas Number : 55377-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52648
Availability details for
Quantity
  • Availability
  • Add to Cart