ENA245485435 Enamine

1-[(2-methoxyphenyl)carbonyl]-1,4-diazepane

  • MDL number MFCD05879251
  • Cas Number : 61903-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52660

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52660
Availability details for
Quantity
  • Availability
  • Add to Cart