ENA245485437 Enamine

2,2,2-trifluoroethyl N-(1,1-dioxo-1lambda6-thiolan-3-yl)carbamate

  • MDL number MFCD13195844
  • Cas Number : 1197746-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52667
Availability details for
Quantity
  • Availability
  • Add to Cart