ENA245485441 Enamine

2,2,2-trifluoroethyl N-(2-methoxyethyl)carbamate

  • MDL number MFCD13195848
  • Cas Number : 1209447-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52676

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52676
Availability details for
Quantity
  • Availability
  • Add to Cart