ENA245485442 Enamine

2,2,2-trifluoroethyl N-[2-(morpholin-4-yl)ethyl]carbamate

  • MDL number MFCD13195849
  • Cas Number : 1197808-29-9
  • Molecular Weight : 256.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-52677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52677
Availability details for
Quantity
  • Availability
  • Add to Cart