ENA245485443 Enamine

2,2,2-trifluoroethyl N-[2-(dimethylamino)-2-(thiophen-2-yl)ethyl]carbamate

  • MDL number MFCD13195850
  • Cas Number : 1197466-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52679
Availability details for
Quantity
  • Availability
  • Add to Cart