ENA245485449 Enamine

N-ethyl-3,5-dimethyl-N-propyl-1H-pyrazole-4-sulfonamide hydrochloride

  • MDL number MFCD12913147
  • Cas Number : 1239733-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52697
Availability details for
Quantity
  • Availability
  • Add to Cart