ENA245485450 Enamine

ethyl({5-[4-(trifluoromethyl)phenyl]furan-2-yl}methyl)amine

  • MDL number MFCD12691269
  • Cas Number : 1153421-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52698
Availability details for
Quantity
  • Availability
  • Add to Cart